Imprimer
Chemical resistant overalls, Microchem® 3000
Fournisseur: Ansell
YE30W-00111-02 YE30W-00111-03 YE30W-00111-04 YE30W-00111-05 YE30W-00111-06 YE30W-00111-07
114-9503EA 0 EUR
114-9503 114-9504 114-9505 114-9506 114-9507 114-9508
Chemical resistant overalls, Microchem® 3000
Vêtements à usage unique
These overalls are manufactured from one of the lightest and most comfortable chemical protective materials on the market. This durable multi-layer fabric (two non woven layers, one external film barrier layer) provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Two-piece hood, double zip closure, double cuffs
  • Elasticated hood, waist, double cuffs and ankles
  • Excellent liquid penetration resistance and barrier to fine particulates (>0,01 µm)
  • Moisture vapour permeable ('breathable') to help reduce the risk of heat stress
  • Silicone free, low-linting
  • Ultrasonically welded seams provide a strong liquid and particle barrier
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, mining.
Certifications: Type 3-B/4-B/5-B, 1149-5, EN 14126, EN 1073-2. Suitable for Ex-Zones.