Imprimer
Scalpel blades, stainless steel, Feather®
Fournisseur: PFM Medical GmbH
201000024 201000012 201000015 02.01000023
FEAT201000024EA 25.7 EUR
FEAT201000024 FEAT201000012 FEAT201000015 FEAT02.01000023
Scalpel blades, stainless steel, Feather®
Bistouris et pièces détachées Lames de bistouri
These stainless steel blades made from the highest quality hardened stainless steel are suitable for all surgical applications.
  • Outstanding quality and performance thanks to optimised micrometer-accurate precision grinding
  • Double-honing process ensures ultra-sharp cutting edges of great durability
  • Ready for use immediately after removal from the package
  • Balanced design
Conditionnement: Blades are individually sterile packed. 100 blades per pack.