Imprimer
Capillary columns, Rtx®-440 (fused silica)
Fournisseur: Restek
12923 12924 12938 12939 12940 42902
REST12923EA 0 EUR
REST12923 REST12924 REST12938 REST12939 REST12940 REST42902
Capillary columns, Rtx®-440 (fused silica)
Colonnes pour chromatographie
These general purpose capillary columns with unique selectivity for pesticides, PAHs, or other semivolatiles are ideal for low/trace-level analyses.
  • Midpolarity proprietary Crossbond phase
  • Low-bleed, high-resolution columns with unique selectivity
  • Wide temperature range from 20 to 340 °C