Imprimer
Anti-PTGDS Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
NB110-59911 NB110-59911SS
NOVUNB110-59911EA 606 EUR
NOVUNB110-59911 NOVUNB110-59911SS
Anti-PTGDS Rabbit Polyclonal Antibody
Anticorps
Rabbit Polyclonal Prostaglandin D Synthase/Lipocalin Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Feline, Primate, Rabbit.
Type: Primary
Antigen: Prostaglandin D Synthase/Lipocalin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human