Imprimer
Anti-NGFR Mouse Monoclonal Antibody (CF405S) [clone: NGFR5 NTR/912]
Fournisseur: Biotium
BNC040914-500 BNC040914-100
BNC040914-500EA 560 EUR
BNC040914-500 BNC040914-100
Anti-NGFR Mouse Monoclonal Antibody (CF405S) [clone: NGFR5 NTR/912]
Anticorps
This antibody recognizes a glycoprotein of 75 kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells, and myoepithelial cells. Anti-NGFR stains the myoepithelial cells of breast ducts and intra-lobular fibroblasts of breast ducts.
Type: Primary
Antigen: NGFR
Clonality: Monoclonal
Clone: NGFR5 NTR/912
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Non-human primates