Imprimer

Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: HSPD1/780]

Fournisseur: Biotium
BNC040780-500 BNC040780-100
BTIUBNC040780-500EA 560 EUR
BTIUBNC040780-500 BNC040780-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: HSPD1/780]
Anticorps
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: HSPD1/780
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Cow, Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR