Imprimer

Anti-HSPD1 Mouse Monoclonal Antibody (CF647) [clone: GROEL/730]

Fournisseur: Biotium
BNC470730-500 BNC470730-100
BTIUBNC470730-500EA 560 EUR
BTIUBNC470730-500 BTIUBNC470730-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF647) [clone: GROEL/730]
Anticorps
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog. Clone GROEL730 reacts only with human and is useful in distinguishing human hsp60 from other mammalian and bacterial hsp60.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: GROEL/730
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR