Imprimer
Acide citrique monohydraté, powder_white crystalline
Fournisseur: MP Biomedicals
Attention
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
02150699.5 0215069901 0215069905
ICNA02150699.5EA 45.6 EUR
ICNA02150699.5 ICNA0215069901 ICNA0215069905
Acide citrique monohydraté, powder_white crystalline
Acide citrique monohydraté
Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilization of nutritional calcium. It is a useful buffer component.
Soluble in water, methanol.
Formule: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Point d'éclair: 173,9
Numéro MDL: MFCD00149972
Numéro CAS: 5949-29-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM