Imprimer
L(+)-Leucine, poudre blanche
Fournisseur: MP Biomedicals
Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
02102158.3 0210215805 02102158.1 0210215801
ICNA02102158.3EA 25 EUR
ICNA02102158.3 ICNA0210215805 ICNA02102158.1 ICNA0210215801
L(+)-Leucine, poudre blanche
L(+)-Leucine
Leucine is a branched-chain α-amino acid. Leucine is classified as a hydrophobic amino acid due to its aliphatic isobutyl side chain.
Leucine has been used as a molecular marker in the recovery of DNA from palaeontological samples for PCR analysis.
Soluble in water (25 mg/ml); sparingly soluble in alcohol.
Attention: Research Use Only (RUO).
Formule: C₆H₁₃NO₂
MW: 131.17 g/mol
Point de fusion: 293 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00002617
Numéro CAS: 61-90-5
Merck Index: 14,05451
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
FTIR Conforms to Standard
Specific Rotation +14,5 ° to +16,5 °
Loss on drying ≤0.2%