Imprimer
Capillary columns, DB-210
Fournisseur: Agilent
122-0212 122-0232 122-0233 123-0213 123-0232 123-0233 125-0232
HEWL122-0212EA 455 EUR
HEWL122-0212 HEWL122-0232 HEWL122-0233 HEWL123-0213 HEWL123-0232 HEWL123-0233 HEWL125-0232
Capillary columns, DB-210
Colonnes pour chromatographie
These (50%-Trifluoropropyl)-methylpolysiloxane columns are excellent for U.S. EPA Methods 8140 and 609.
  • High polarity
  • Bonded and cross-linked
  • Solvent rinsable
  • Exact replacement of HP-210
  • Close equivalent to USP Phase G6