Imprimer
Safety shoes, lace-up, uvex quatro pro safe, 8400
Fournisseur: Uvex
uvex quatro pro safe
114-7055EA 134 EUR
114-7055 114-7056 114-7053 114-7054 114-7059 114-7048 114-7049 114-7057 114-7058 114-7051 114-7052 114-7060 114-7050
Safety shoes, lace-up, uvex quatro pro safe, 8400
Chaussures et bottes
Sturdy, comfortable anti-static safety shoe made from water resistant leather upper, soft padded collar and padded dust tongue plus a dual-density, self-cleaning, slip resistant PUR sole (4,5 mm, heat resistant to +120 °C, oil and petrol resistant also silicone-free).
  • Climazone technology for improved comfort
  • Breathable textile lining
  • Removable, anti-static insole with a moisture control system creates a superior foot climate and improved comfort
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Sole offers integrated heel support
  • Smooth running, dirt resistant lacing system
  • Foam scuff cap for additional protection
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifications: DGUV 112-191; EN ISO 20345:2011; S3, SRC