Imprimer
Bisphenol A diglycidyl ether resin
Fournisseur: Thermo Fisher Scientific
Attention
L14817.14 L14817.22 L14817.36
L14817.14EA 30.6 EUR
L14817.14 L14817.22 L14817.36
Bisphenol A diglycidyl ether resin
Bisphenol A diglycidyl ether resin
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Formule: C₂₁H₂₄O₄
MW: 340.42 g/mol
Densité: 1.17
Numéro MDL: MFCD00080480
Numéro CAS: 1675-54-3
EINECS: 216-823-5
ADR: 9 III