Imprimer
DL-Asparagine monohydrate 98%
Fournisseur: Thermo Fisher Scientific
Synonyms: DL-Aspartic acid 4-amide monohydrate, H-DL-Asn-OH monohydrate, (±)-2-Aminosuccinic acid 4-amide monohydrate, DL-Asparagine hydrate, H-DL-Asn-OH·H₂O, DL-2-Aminosuccinamic acid monohydrate, 2,4-Diamino-4-oxobutanoic acid monohydrate
B20273.22 B20273.36
B20273.22EA 86.3 EUR
B20273.22 B20273.36
DL-Asparagine monohydrate 98%
DL-Asparagine monohydrate
Purity: 98% CAS No.: 3130-87-8 Molecular Formula: C4H8N2O3.H2O Formula Weight: 150.14 (132.12anhy) Melting Point: ca 220° dec. Beilstein Reference: 6234992 MDL No.: MFCD00151039
Formule: C₄H₈N₂O₃·H₂O
MW: 150.13 g/mol
Point de fusion: ∼ 220 °C (déc.)
Température de stockage: Température ambiante
Numéro MDL: MFCD00151039
Numéro CAS: 69833-18-7
EINECS: 221-521-1