Imprimer
Palladium 5% on activated carbon paste (50% water) 5R39
Fournisseur: Thermo Fisher Scientific
44337.06 44337.14 44337.22
44337.06EA 92.9 EUR
44337.06 44337.14 44337.22
Palladium 5% on activated carbon paste (50% water) 5R39
Palladium
CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879
Applications: Debenzylation, C-N and C-O cleavage, alkene hydrogenation
Formule: Pd
MW: 106.420 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00011167
Numéro CAS: 7440-05-3
EINECS: 231-115-6