Imprimer
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6000
Fournisseur: Thermo Fisher Scientific
42501.AD 42501.AK 42501.K2
42501.ADEA 41.3 EUR
42501.AD 42501.AK 42501.K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6000
Polydimethylsiloxane, trimethylsiloxy terminated
CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 315°(599°F) Physical Form: Viscous liquid Density: 0.950 Refractive Index: 1.4025 Appearance: Colourless Odour: Odourless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360
Formule: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Point d'ébullition: >205 °C
Point de fusion: < –40 °C
Densité: 0.950
Point d'éclair: 315 °C (599 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00148360
Numéro CAS: 9016-00-6