Imprimer
Dual column direct injection tee kit
Fournisseur: Restek
20411 20412
REST20411EA 0 EUR
REST20411 REST20412
Dual column direct injection tee kit
Raccords et connecteurs
The tee is designed with a glass spiral to promote sample vaporisation and to provide even sample distribution to the two columns. The glass spiral also traps sample residue and minimises the need for guard columns. A Press-Tight® taper in each outlet leg facilitates a perfect dead volume free connection to each analytical column and allows visual confirmation of the column connection.
Information de livraison: The kit is delivered with all fittings and ferrules.