Imprimer
Reagent bottles, wide neck, with screw cap, Corning®
Fournisseur: Corning
1500P-250 1500P-2L 1500P-1L 1500P-500
CORN1500P-250EA 94.96 EUR
CORN1500P-250 CORN1500P-2L CORN1500P-1L CORN1500P-500
Reagent bottles, wide neck, with screw cap, Corning®
Flacons Flacons en plastique
PP, translucent, with PP screw cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
Can be used anywhere in the laboratory, as well as for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.