Imprimer
Digital calipers
Fournisseur: MOINEAU
1150MI 1152MI
MOIN1150MIEA 82.9 EUR
MOIN1150MI MOIN1152MI
Digital calipers
Pieds à coulisse
Digital calipers with crossed steps.
  • Made of chromed stainless steel
  • Large display with automatic switch-off
  • Self-locking instrument
Weight: 250 g
Information de livraison: Supplied in a box with an included battery and a spare battery.