Imprimer
Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: 10F723]
Fournisseur: US Biological
H1830-95M-100 H1830-95M-25
USBIH1830-95M-100EA 608 EUR
USBIH1830-95M-100 USBIH1830-95M-25
Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: 10F723]
Anticorps
Recommended dilution: Western Blot:1:5000; Immunochemistry:1:500
Type: Primary
Antigen: Heat Shock Protein 70
Clonality: Monoclonal
Clone: 10F723
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat,