Imprimer
Safety shoes, lace-up, Josio
Fournisseur: PARADE
JOSIO6804/39 JOSIO6804/40 JOSIO6804/41 JOSIO6804/42 JOSIO6804/45 JOSIO6804/43 JOSIO6804/44 JOSIO6804/46 JOSIO6804/47 JOSIO6804/35 JOSIO6804/36 JOSIO6804/37 JOSIO6804/38
PARAJOSIO6804/39EA 0 EUR
PARAJOSIO6804/39 PARAJOSIO6804/40 PARAJOSIO6804/41 PARAJOSIO6804/42 PARAJOSIO6804/45 PARAJOSIO6804/43 PARAJOSIO6804/44 PARAJOSIO6804/46 PARAJOSIO6804/47 PARAJOSIO6804/35 PARAJOSIO6804/36 PARAJOSIO6804/37 PARAJOSIO6804/38
Safety shoes, lace-up, Josio
Chaussures et bottes
Lace-up safety shoes made from full grain leather upper and a dual-density PU sole.
  • Anti-fungal, antibacterial, anti-presperation polyamide lining
  • Protective steel toecap (200 J)
  • Temperature resistant over 100 °C
Colour: Black
Colour: Black
Certifications: EN 20345; S2 SRC