Imprimer

Anti-6xHis Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Rockland Immunochemicals
ROCK600-406-382EA 706 EUR
ROCK600-406-382
Anti-6xHis Rabbit Polyclonal Antibody (Biotin)
Anticorps
Primary Rabbit Anti-6X HIS EPITOPE TAG Reacts with Pan

Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR