Imprimer

Anti-VSV-G Rabbit Polyclonal Antibody

Fournisseur: Rockland Immunochemicals
ROCK600-401-386EA 504 EUR
ROCK600-401-386
Anti-VSV-G Rabbit Polyclonal Antibody
Anticorps
Primary Rabbit Anti-VSV-G EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: VSV-G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR