Imprimer

Anti-Protein A Goat Polyclonal Antibody (Biotin)

Fournisseur: Rockland Immunochemicals
ROCK200-1677EA 3780 EUR
ROCK200-1677
Anti-Protein A Goat Polyclonal Antibody (Biotin)
Anticorps
Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR