Imprimer

Anti-NDM-1 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
NBP1-77688 NBP1-77688SS
NOVUNBP1-77688EA 591 EUR
NOVUNBP1-77688 NOVUNBP1-77688SS
Anti-NDM-1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NDM-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bacteria
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR