Imprimer

Anti-DGAT2 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
NBP1-71701 NBP1-71701SS
NOVUNBP1-71701EA 568 EUR
NOVUNBP1-71701 NOVUNBP1-71701SS
Anti-DGAT2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR