Imprimer
Anti-SLC7A11 Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
NB300-317 NB300-317SS
NOVUNB300-317EA 647 EUR
NOVUNB300-317 NOVUNB300-317SS
Anti-SLC7A11 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human