Imprimer

Anti-HIST3H3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
NB21-1101 NB21-1101SS
NOVUNB21-1101EA 612 EUR
NOVUNB21-1101 NOVUNB21-1101SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR