Imprimer
Anti-ATG16L1 Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
NB110-60928 NB110-60928SS
NOVUNB110-60928EA 554 EUR
NOVUNB110-60928 NOVUNB110-60928SS
Anti-ATG16L1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant