Imprimer
Anti-IgG Chicken Polyclonal Antibody (CF™ 594)
Fournisseur: Biotium
20223 20223-1
20223.EA 300 EUR
20223. 20223-1
Anti-IgG Chicken Polyclonal Antibody (CF™ 594)
Anticorps
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Rabbit