Imprimer
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Fournisseur: Biotium
20153 20153-1
20153.EA 238 EUR
20153. BTIU20153-1
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Anticorps
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™594 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Deep-red fluorescent CF™594 (Ex/Em 593/614 nm) is spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugate, and has excellent photostability.
Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit