Imprimer
Anti-c-Met Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB99414-100 ORB99414-20
BIRBORB99414-100EA 0 EUR
BIRBORB99414-100 BIRBORB99414-20
Anti-c-Met Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:2000; Immunofluorescence: 1:100-1:500
Type: Primary
Antigen: c-Met
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat