Imprimer

Anti-HSPE1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB67557-100 ORB67557-25
BIRBORB67557-100EA 0 EUR
BIRBORB67557-100 BIRBORB67557-25
Anti-HSPE1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:1000

Type: Primary
Antigen: HSPE1 (heat shock 10kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR