Imprimer

Anti-RBP4 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB136234-100 ORB136234-200 ORB136234-50
BIRBORB136234-100EA 0 EUR
BIRBORB136234-100 BIRBORB136234-200 BIRBORB136234-50
Anti-RBP4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: RBP4 (retinol binding protein 4, plasma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR