Imprimer
Anti-PSMD2 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB128590-100 ORB128590-200
BIRBORB128590-100EA 0 EUR
BIRBORB128590-100 BIRBORB128590-200
Anti-PSMD2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PSMD2 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat