Imprimer
Anti-PPP1R8 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB128562-100 ORB128562-200
BIRBORB128562-100EA 0 EUR
BIRBORB128562-100 BIRBORB128562-200
Anti-PPP1R8 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PPP1R8 (protein phosphatase 1, regulatory subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat