Imprimer

Anti-AKR1A1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB127665-100 ORB127665-200
BIRBORB127665-100EA 0 EUR
BIRBORB127665-100 BIRBORB127665-200
Anti-AKR1A1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR