Imprimer

Anti-MER/TYRO3 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB127574-100 ORB127574-200
BIRBORB127574-100EA 0 EUR
BIRBORB127574-100 BIRBORB127574-200
Anti-MER/TYRO3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MER/TYRO3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR