Imprimer

Anti-CATG Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB126613-100 ORB126613-200
BIRBORB126613-100EA 0 EUR
BIRBORB126613-100 BIRBORB126613-200
Anti-CATG Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CATG
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR