Imprimer
Anti-PEG3 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB126237-100 ORB126237-200
BIRBORB126237-100EA 0 EUR
BIRBORB126237-100 BIRBORB126237-200
Anti-PEG3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PEG3 (paternally expressed 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human