Imprimer

Anti-ALDH3B1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB125413-100 ORB125413-200
BIRBORB125413-100EA 0 EUR
BIRBORB125413-100 BIRBORB125413-200
Anti-ALDH3B1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ALDH3B1 (aldehyde dehydrogenase 3 family, member B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR