Imprimer
Anti-ADH7 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB125392-100 ORB125392-200
BIRBORB125392-100EA 0 EUR
BIRBORB125392-100 BIRBORB125392-200
Anti-ADH7 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ADH7 (alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human