Imprimer

Anti-PARP1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB106557-100 ORB106557-20
BIRBORB106557-100EA 0 EUR
BIRBORB106557-100 BIRBORB106557-20
Anti-PARP1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:2000

Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR