Imprimer

Anti-MAP2K3 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB106449-100 ORB106449-20
BIRBORB106449-100EA 0 EUR
BIRBORB106449-100 BIRBORB106449-20
Anti-MAP2K3 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR