Imprimer

Anti-DCK Rabbit polyclonal antibody

Fournisseur: Biorbyt
ORB101830-200 ORB101830-100
BIRBORB101830-200EA 0 EUR
BIRBORB101830-200 BIRBORB101830-100
Anti-DCK Rabbit polyclonal antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: DCK (deoxycytidine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR