Imprimer

Anti-ALDH1L1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB101820-100 ORB101820-200
BIRBORB101820-100EA 0 EUR
BIRBORB101820-100 BIRBORB101820-200
Anti-ALDH1L1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: ALDH1L1 (aldehyde dehydrogenase 1 family, member L1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR