Imprimer

Anti-PRR11 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB101497-100 ORB101497-200
BIRBORB101497-100EA 0 EUR
BIRBORB101497-100 BIRBORB101497-200
Anti-PRR11 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PRR11 (proline rich 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR