Imprimer

Anti-HSPE1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB101345-100 ORB101345-200
BIRBORB101345-100EA 0 EUR
BIRBORB101345-100 BIRBORB101345-200
Anti-HSPE1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HSPE1 (heat shock 10kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR