Imprimer

Anti-PIP Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB101264-100 ORB101264-200
BIRBORB101264-100EA 0 EUR
BIRBORB101264-100 BIRBORB101264-200
Anti-PIP Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PIP (prolactin-induced protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR