Imprimer
Anti-KL Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
ORB101064-100 ORB101064-200
BIRBORB101064-100EA 0 EUR
BIRBORB101064-100 BIRBORB101064-200
Anti-KL Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: KL (klotho)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant