Imprimer

Anti-pan methyl Lysine Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
ORB100017-100 ORB100017-200
BIRBORB100017-100EA 0 EUR
BIRBORB100017-100 BIRBORB100017-200
Anti-pan methyl Lysine Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: pan methyl Lysine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR